COM448687722 Combi-Blocks, Inc.

8-bromo-5-fluoro-4-hydroxyquinoline-2-carboxylic acid

  • MDL number MFCD27578981
  • Cas Number : 1696191-67-9
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5397
Availability details for
Quantity
  • Availability
  • Add to Cart