COM448687728 Combi-Blocks, Inc.

2-Amino-4-cyclopropylpyrimidine-5-carboxylic acid

  • MDL number MFCD16082818
  • Cas Number : 1247083-73-3
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5412
Availability details for
Quantity
  • Availability
  • Add to Cart