COM448687731 Combi-Blocks, Inc.

2-Amino-4-(3-chlorophenyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD06850048
  • Cas Number : 899349-06-5
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5421
Availability details for
Quantity
  • Availability
  • Add to Cart