COM448687736 Combi-Blocks, Inc.

8-Methoxy-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid

  • MDL number MFCD09945551
  • Cas Number : 93445-77-3
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5432
Availability details for
Quantity
  • Availability
  • Add to Cart