COM448687739 Combi-Blocks, Inc.

2-amino-4-(4-fluorophenyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD27578988
  • Cas Number : 1820665-87-9
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5436
Availability details for
Quantity
  • Availability
  • Add to Cart