COM448687740 Combi-Blocks, Inc.

6,8-Dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid

  • MDL number MFCD18343448
  • Cas Number : 134785-79-8
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5442
Availability details for
Quantity
  • Availability
  • Add to Cart