COM448687743 Combi-Blocks, Inc.

5-tert-Butyl-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD24039267
  • Cas Number : 1152540-42-5
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5453
Availability details for
Quantity
  • Availability
  • Add to Cart