COM448687744 Combi-Blocks, Inc.

4,8-Dichloroquinoline-2-carboxylic acid

  • MDL number MFCD06810558
  • Cas Number : 902742-81-8
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5454
Availability details for
Quantity
  • Availability
  • Add to Cart