COM448687745 Combi-Blocks, Inc.

5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD27578990
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5455
Availability details for
Quantity
  • Availability
  • Add to Cart