COM448687751 Combi-Blocks, Inc.

4-chloro-6-acetamidoquinoline-2-carboxylic acid

  • MDL number MFCD27578992
  • Cas Number : 1820711-03-2
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5467

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5467
Availability details for
Quantity
  • Availability
  • Add to Cart