COM448687755 Combi-Blocks, Inc.

6,8-Difluoro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid

  • MDL number MFCD18343494
  • Cas Number : 1267474-18-9
  • Molecular Weight : 225.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5476
Availability details for
Quantity
  • Availability
  • Add to Cart