COM448687762 Combi-Blocks, Inc.

4,5-Difluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number MFCD24039806
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5484
Availability details for
Quantity
  • Availability
  • Add to Cart