COM448687764 Combi-Blocks, Inc.

5-Bromo-4-chloro-8-methylquinoline-2-carboxylic acid

  • MDL number MFCD18343507
  • Cas Number : 1340075-78-6
  • Molecular Weight : 300.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5490
Availability details for
Quantity
  • Availability
  • Add to Cart