COM448687765 Combi-Blocks, Inc.

2-Quinolinecarboxylic acid, 4-chloro-6-fluoro-

  • MDL number MFCD06810565
  • Cas Number : 902742-88-5
  • Molecular Weight : 225.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5491

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5491
Availability details for
Quantity
  • Availability
  • Add to Cart