COM448687770 Combi-Blocks, Inc.

2-Methyl-4-(3-chloro)phenyl thiazole-5-carboxylic acid

  • MDL number MFCD08458376
  • Cas Number : 905808-17-5
  • Molecular Weight : 253.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5506

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5506
Availability details for
Quantity
  • Availability
  • Add to Cart