COM448687774 Combi-Blocks, Inc.

5-tert-butyl-1-(6-chloropyridin-2-yl)pyrazole-4-carboxylic acid

  • MDL number MFCD27578999
  • Cas Number : 1820740-45-1
  • Molecular Weight : 279.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5518

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5518
Availability details for
Quantity
  • Availability
  • Add to Cart