COM448687776 Combi-Blocks, Inc.

5-Cyclopropyl-1-(pyridin-2-yl)-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD11156671
  • Cas Number : 1152975-89-7
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5521
Availability details for
Quantity
  • Availability
  • Add to Cart