COM448687783 Combi-Blocks, Inc.

5-P-Tolyl-1h-pyrazole-4-carboxylic acid

  • MDL number MFCD07357449
  • Cas Number : 879996-66-4
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5532
Availability details for
Quantity
  • Availability
  • Add to Cart