COM448687788 Combi-Blocks, Inc.

5-methyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxylic acid

  • MDL number MFCD27579004
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5551
Availability details for
Quantity
  • Availability
  • Add to Cart