COM448687790 Combi-Blocks, Inc.

2-Amino-4-(4-chlorophenyl)-5-thiazolecarboxylic acid

  • MDL number MFCD06849882
  • Cas Number : 206555-62-6
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5554
Availability details for
Quantity
  • Availability
  • Add to Cart