COM448687795 Combi-Blocks, Inc.

3-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzoic acid

  • MDL number MFCD27579007
  • Cas Number : 2377608-22-3
  • Molecular Weight : 324.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5570
Availability details for
Quantity
  • Availability
  • Add to Cart