COM448687796 Combi-Blocks, Inc.

(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid hydrate

  • MDL number MFCD27579008
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5574
Availability details for
Quantity
  • Availability
  • Add to Cart