COM448687803 Combi-Blocks, Inc.

Beta-alanine hydrochloride

  • MDL number MFCD27579009
  • Cas Number : 6057-90-5
  • Molecular Weight : 125.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5589
Availability details for
Quantity
  • Availability
  • Add to Cart