COM448687806 Combi-Blocks, Inc.

Aminoisobutyric acid hydrochloride

  • MDL number MFCD22385038
  • Cas Number : 5938-34-1
  • Molecular Weight : 139.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5593
Availability details for
Quantity
  • Availability
  • Add to Cart