COM448687807 Combi-Blocks, Inc.

D-methionine hydrochloride

  • MDL number MFCD27579011
  • Cas Number : 84228-57-9
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5594

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5594
Availability details for
Quantity
  • Availability
  • Add to Cart