COM448687808 Combi-Blocks, Inc.

(S)-2-Amino-3-(4-nitrophenyl)propanoic acid hydrochloride

  • MDL number MFCD21604237
  • Cas Number : 906813-62-5
  • Molecular Weight : 246.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5598
Availability details for
Quantity
  • Availability
  • Add to Cart