COM448687811 Combi-Blocks, Inc.

L-tert-Leucine hydrochloride

  • MDL number MFCD07368368
  • Cas Number : 139163-43-2
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5605

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5605
Availability details for
Quantity
  • Availability
  • Add to Cart