COM448687812 Combi-Blocks, Inc.

D-Phenylalanine, HCl

  • MDL number MFCD12910697
  • Cas Number : 28069-46-7
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5608

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5608
Availability details for
Quantity
  • Availability
  • Add to Cart