COM448687813 Combi-Blocks, Inc.

L-tryptophan hydrochloride

  • MDL number MFCD27579013
  • Cas Number : 6159-33-7
  • Molecular Weight : 240.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5609
Availability details for
Quantity
  • Availability
  • Add to Cart