COM448687814 Combi-Blocks, Inc.

valine hydrochloride

  • MDL number MFCD27579014
  • Cas Number : 25616-14-2
  • Molecular Weight : 153.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5611
Availability details for
Quantity
  • Availability
  • Add to Cart