COM448687816 Combi-Blocks, Inc.

4-Bromo-l-phe-oh hcl

  • MDL number MFCD09028124
  • Cas Number : 122839-59-2
  • Molecular Weight : 280.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5623
Availability details for
Quantity
  • Availability
  • Add to Cart