COM448687817 Combi-Blocks, Inc.

5-(aminomethyl)-2-methoxybenzoic acid hydrochloride

  • MDL number MFCD27579015
  • Cas Number : 1820666-91-8
  • Molecular Weight : 217.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5628

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5628
Availability details for
Quantity
  • Availability
  • Add to Cart