COM448687819 Combi-Blocks, Inc.

benzylcysteine hydrochloride

  • MDL number MFCD27579017
  • Cas Number : 103967-49-3
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5633
Availability details for
Quantity
  • Availability
  • Add to Cart