COM448687821 Combi-Blocks, Inc.

L-arginine dihydrochloride

  • MDL number MFCD27579018
  • Cas Number : 58107-60-1
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5638
Availability details for
Quantity
  • Availability
  • Add to Cart