COM448687829 Combi-Blocks, Inc.

2-[2-(Methanesulfonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD18734138
  • Cas Number : 2377611-00-0
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0008
Availability details for
Quantity
  • Availability
  • Add to Cart