COM448687831 Combi-Blocks, Inc.

2,6-Difluorobenzylmethylsulfone

  • MDL number MFCD06798025
  • Cas Number : 886497-97-8
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0010

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0010
Availability details for
Quantity
  • Availability
  • Add to Cart