COM448687833 Combi-Blocks, Inc.

Methylsulfonyldiphenylmethane

  • MDL number MFCD00047809
  • Cas Number : 25195-40-8
  • Molecular Weight : 246.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0012

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0012
Availability details for
Quantity
  • Availability
  • Add to Cart