COM448687837 Combi-Blocks, Inc.

3,5-Bis(trifluoromethyl)benzylmethylsulfone

  • MDL number MFCD06798028
  • Cas Number : 261924-41-8
  • Molecular Weight : 306.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0016

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0016
Availability details for
Quantity
  • Availability
  • Add to Cart