COM448687839 Combi-Blocks, Inc.

4-Chloro-2-fluorobenzylmethylsulfone

  • MDL number MFCD06798037
  • Cas Number : 25195-55-5
  • Molecular Weight : 222.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0018

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0018
Availability details for
Quantity
  • Availability
  • Add to Cart