COM448687840 Combi-Blocks, Inc.

3-Trifluoromethylbenzylmethylsulfone

  • MDL number MFCD06798013
  • Cas Number : 25195-49-7
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0019
Availability details for
Quantity
  • Availability
  • Add to Cart