COM448687841 Combi-Blocks, Inc.

4-(Methanesulfonylmethyl)-n-methylbenzamide

  • MDL number MFCD09347002
  • Cas Number : 930939-62-1
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0020
Availability details for
Quantity
  • Availability
  • Add to Cart