COM448687844 Combi-Blocks, Inc.

1-[3-(Methanesulfonylmethyl)-4-methoxyphenyl]ethan-1-one

  • MDL number MFCD16664697
  • Cas Number : 1249404-09-8
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0023
Availability details for
Quantity
  • Availability
  • Add to Cart