COM448687848 Combi-Blocks, Inc.

2-Methanesulfonyl-2-phenylacetic acid

  • MDL number MFCD24482578
  • Cas Number : 29270-21-1
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0027
Availability details for
Quantity
  • Availability
  • Add to Cart