COM448687850 Combi-Blocks, Inc.

2-Fluorobenzylmethylsulfone

  • MDL number MFCD06798006
  • Cas Number : 25195-45-3
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0029
Availability details for
Quantity
  • Availability
  • Add to Cart