COM448687852 Combi-Blocks, Inc.

3-(Methylsulfonylmethyl)phenylboronic acid pinacol ester

  • MDL number MFCD18734144
  • Cas Number : 1864802-09-4
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0031
Availability details for
Quantity
  • Availability
  • Add to Cart