COM448687853 Combi-Blocks, Inc.

N-tert-Butyl-4-(methanesulfonylmethyl)benzamide

  • MDL number MFCD07949689
  • Cas Number : 1050648-37-7
  • Molecular Weight : 269.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0032
Availability details for
Quantity
  • Availability
  • Add to Cart