COM448687854 Combi-Blocks, Inc.

4-(Methanesulfonylmethyl)-n-(propan-2-yl)benzamide

  • MDL number MFCD11717016
  • Cas Number : 1089613-97-7
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0033
Availability details for
Quantity
  • Availability
  • Add to Cart