COM448687859 Combi-Blocks, Inc.

N-Ethyl-4-(methanesulfonylmethyl)benzamide

  • MDL number MFCD09351850
  • Cas Number : 930955-08-1
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0038
Availability details for
Quantity
  • Availability
  • Add to Cart