COM448687860 Combi-Blocks, Inc.

1,3,2-Dioxaborolane, 2-[3-fluoro-4-[(methylsulfonyl)methyl]phenyl]-4,4,5,5-tetramethyl-

  • MDL number MFCD18760488
  • Cas Number : 1351499-67-6
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0039

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0039
Availability details for
Quantity
  • Availability
  • Add to Cart