COM448687863 Combi-Blocks, Inc.

2-Bromo-4-fluoro-1-methyl-3-((methylsulfonyl)methyl)benzene

  • MDL number MFCD22987924
  • Cas Number : 1820614-09-2
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0042

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YE-0042
Availability details for
Quantity
  • Availability
  • Add to Cart